Learn More
IBA GmbH™ Nylon Filter Tubes for Streptamer™ Cell Isolation
IBA GmbH™ Nylon Filter Tubes for Streptamer™ Cell Isolation can be used for a variety of applications like removal of cell clumps and debris.
Brand:  IBA GmbH™ 6-5601-010
Additional Details : Weight : 0.00200kg
Product Code. 15673888
  • 54.00€ / Pack of 10
Estimated Shipment date
from Supplier 09-09-2019
Add to basket
Description
Description
  • Suitable for removal of cell clumps and debris
  • Can be used to obtain single-cell suspensions
  • Protects flow cytometry and cell sorting instrumentation
  • Non-sterile
Specifications
Specifications
Nylon
35μm mesh size