Learn More
IBA GmbH™ Nylon Filter Tubes for Streptamer™ Cell Isolation
IBA GmbH™ Nylon Filter Tubes for Streptamer™ Cell Isolation can be used for a variety of applications like removal of cell clumps and debris.
Brand: IBA GmbH™ 6-5601-010
Additional Details : Weight : 0.00200kg
Description
- Suitable for removal of cell clumps and debris
- Can be used to obtain single-cell suspensions
- Protects flow cytometry and cell sorting instrumentation
- Non-sterile
Specifications
| Nylon | |
| 35μm mesh size |