Learn More
Macherey Nagel Bioanalysis™ Nucleospin™ 8 Plant Ii Core Kit
Rapid isolation of DNA from a multitude of plant samples in flexible 8‐well strip format
Brand:  Macherey Nagel Bioanalysis™ 740467.4
Product Code. 12748432
  • 933.00€ / 384 reactions
Estimated Shipment: 03-01-2025
Description
contains Plant II binding strips, buffers, RNase A, 48 x 8 preps Macherey-Nagel
Specifications
Specifications
18°C to 25°C
Plants
384 Preps
50 bp–approx. 50 kbp
8-well Strip
Isolation of DNA
20–100 mg wet weight
DNA
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1