Learn More
WTW™ Nitrate Test Kits
Product Code. 11734774
Click to view available options
Range:
0.2 to 13.0 mg/L NO3-N, 0.9 to 57.5 mg/L NO3
0.2 to 20 mg/L NO3-N, 0.9 to 88.5 mg/L NO3
0.5 to 18 mg/L NO3-N, 2.2 to 79.7 mg/L NO3
0.5 to 25 mg/L NO3-N, 2.2 to 110.7 mg/L NO3
1 to 50 mg/L NO3-N, 4 to 221 mg/L NO3
No. of Tests:
25
50
60
90
Unit Size:
0.52 L
1 set
300g
Pack of 25
Product Code. 11734774
Brand: WTW™ 250410
Obtain precision measurements for waste water, drinking water, and surface water with CombiCheck Nitrate (NO3) cell test kits.
  • Variety of instrumentation for measuring nitrates
  • For waste water, drinking water, surface water
  • PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
  • CombiCheck Nitrate (NO<sub>3</sub>) cell test kit
TRUSTED_SUSTAINABILITY
Specifications
Analytes Nitrospectral
For Use With (Application) For waste water, drinking water, surface water
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Includes Certificates
No. of Tests 60
Range 0.5 to 18 mg/L NO3-N, 2.2 to 79.7 mg/L NO3
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck Nitrate (NO3) Cell Test Kit
compliance-icons
Product Identifier
  • Nitrate Cell Test Method: photometric 0.5 - 18.0 mg/l NO
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid