missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Nicotinamide Adenine Dinucleotide Phosphate, Monosodium Salt, Tetrahydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Plastic bottle
Specifications
Physical Form | Powder |
Quantity | 5 g |
Beilstein | 4779954 |
Formula Weight | 837.45 |
Packaging | Plastic bottle |
Chemical Name or Material | beta-Nicotinamide adenine dinucleotide phosphate monosodium salt tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'