missing translation for 'onlineSavingsMsg'
Learn More

Nickel(Ii) Sulfate Hexahydrate, ACS Reagent, Honeywell Fluka™

Catalog Number 15622260
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
10101-97-0
H12NiO10S
262.839
MFCD00149813
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
This item is not returnable. View return policy
10101-97-0
H12NiO10S
262.839
MFCD00149813
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
5284429
CHEBI:53437
nickel(2+);sulfate;hexahydrate
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 2.5 kg
Linear Formula NiSO4 · 6H2O
UN Number 3077
Formula Weight 262.85g/mol
Percent Purity 99%
Grade ACS Reagent
Density Vapor Density: 2.07 (vs air)
Chemical Name or Material Nickel(Ii) Sulfate Hexahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'