missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NGI 1

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
NGI 1 is an oligosaccharyltransferase (OST) inhibitor; anti-flaviviral. Inhibits dengue, Zika, West Nile, and yellow fever viruses by blocking viral RNA replication (EC50 values are 0.85 and 2.2 μM for DENV and ZIKV inhibition, respectively). Selectively arrests proliferation of non-small-cell lung cancer cells that are dependent on EGFR or FGFR for survival.
Specifications
Specifications
| Chemical Name or Material | 5-[(Dimethylamino)sulfonyl]-N-(5-methyl-2-thiazolyl)-2-(1-pyrrolidinyl)benzamide |
| CAS | 790702-57-7 |
| Quantity | 25 mg |
| Target | Other Transferase Inhibitors |
| Molecular Formula | C17H22N4O3S2 |
| Formula Weight | 394.51 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction