Learn More
Ohaus™ Navigator Portable Balance
Designed for a wide range of industrial, food and laboratory weighing applications. Ohaus™ Navigator NV Balances feature ultra-fast response time and superior overload protection.
202.98€ - 295.80€
Specifications
Material ABS housing
Electrical Requirements 12VDC 0.42A AC adapter (included), 100 to 240V 0.2
Connection Type RS-232, USB or Ethernet Interface
Display Type LCD, 6-digit 7-segment with white LED backlight
Certifications/Compliance OIML F2 or ASTM Class 4
View More Specs
Products 20
Product Code Brand Model Capacity Readability Price Quantity & Availability  
Product Code Brand Model Capacity Readability Price Quantity & Availability  
11742028 View accessory products Ohaus™
83997654
NVT1601 1.6 kg 0.1 g 205.00€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
12797206 Ohaus™
83032042
NVT16000M 16 kg 5 g 240.00€
Each
Estimated Shipment date
from Supplier 04-02-2019
Add to basket
12777206 Ohaus™
83032038
NVT6400M 6.4 kg 2 g 240.00€
Each
Estimated Shipment date
from Supplier 04-02-2019
Add to basket
12747206 Ohaus™
83032030
NVT1601M 1.6 kg 0.5 g 242.76€
Each
Estimated Shipment date
from Supplier 14-02-2019
Add to basket
12717206 Ohaus™
83032304
NV3100M 3.1 kg 1 g 295.80€
Each
Estimated Shipment date
from Supplier 06-02-2019
Add to basket
11979886 Ohaus™
83032034
NVT3200M 3.2 kg 1 g 240.00€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11894353 View accessory products Ohaus™
83997674
NVL20000 20 kg 1 g 270.30€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11874353 View accessory products Ohaus™
83997672
NVL10000 10 kg 1 g 239.70€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11864353 View accessory products Ohaus™
83997670
NVL5101 5.1 kg 0.5 g 239.70€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11824353 View accessory products Ohaus™
83997666
NVL1101 1.1 kg 0.1 g 244.80€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11864343 View accessory products Ohaus™
83997684
NV4101 4.1 kg 0.2 g 229.50€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11804353 View accessory products Ohaus™
83997664
NVL511 510 g 0.1 g 224.40€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11844343 View accessory products Ohaus™
83997682
NV2101 2.1 kg 0.1 g 234.60€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11824343 View accessory products Ohaus™
83997680
NV1101 1.1 kg 0.1 g 224.40€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11804343 View accessory products Ohaus™
83997678
NV511 510 g 0.1 g 202.98€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11782028 View accessory products Ohaus™
83997662
NVT16000 16 kg 1 g 205.00€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11772028 View accessory products Ohaus™
83997660
NVT10001 10 kg 0.5 g 205.00€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11762028 View accessory products Ohaus™
83997658
NVT6401 6.4 kg 0.5 g 205.00€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11752028 View accessory products Ohaus™
83997656
NVT3201 3.2 kg 0.2 g 205.00€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
11884333 View accessory products Ohaus™
83997676
NV212 210 g 0.01 g 244.80€
Each
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
Description
Description
Touchless sensors control the tare, zero, print, function and display backlight minimising cross contamination. A polished stainless steel pan, corrosion free plastic housing and spill resistant pan sub assembly is easy to clean and the optional full body in use cover wraps over and around the housing and keypad to guard against spills, dust and contamination. An optional internal rechargeable lead acid battery can operate LCD versions up to 100 hours (or over twelve 8 hour days) of normal use on a full charge. The rechargeable battery also helps minimise battery replacement and maintenance costs.
  • Designed with two touchless sensors
  • Accurately stabilises in less than one second to provide fast and accurate weighing results
  • Superior overload protection
  • LCD or LED display options
  • Electrical supply 12V d. c. , 0.42A a. c. adapter (included), 100-240V, 0.25A, 50/60Hz Multi-purpose, touch free portable balance for use in a wide range of use in industrial, food and laboratory weighing applications.
Specifications
Specifications
ABS housing
RS-232, USB or Ethernet Interface
OIML F2 or ASTM Class 4
12VDC 0.42A AC adapter (included), 100 to 240V 0.2
LCD, 6-digit 7-segment with white LED backlight