Learn More
Ark Plas Products™ Natural Kynar Premium Grade Barbed Elbow Connectors
An excellent option for connecting tubing in the laboratory, with a pressure rating of up to 125 P.S.I. without clamps.
Brand:  Ark Plas Products™ MCX19-DF0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11891140
  • 45.40€ / Pack of 10
Estimated Shipment
08-04-2021
Add to basket
Description
Description
  • Molded from virgin, lot-traceable material only
  • Manufactured to the most exacting standards
  • Made with critical use applications in mind
  • No parting line on barb
Specifications
Specifications
Natural
Barbed Elbow Connector
5/32 x 5/32 in. Barb
Kynar
10 Pack