Learn More
Gravis Narrow Neck Amber Glass Bottles
Click to view available options
Capacity (Metric):
10 mL
15 mL
250 mL
1090 mL
No. per Pack:
14Pack
195Pack
216Pack
60Pack
Unit Size:
Pack of 14
Pack of 195
Pack of 216
Pack of 60
Description
- Available in a range of capacities
- Manufactured from soda lime glass
- Caps not included
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Amber |
| Diameter (Metric) Inner | 20 mm |
| Material | Soda Lime Glass |
| No. per Pack | 60Pack |
| Mouth | Narrow |
| Height (Metric) | 143 mm |
| Product Type | Amber Bottle |
| Diameter (Metric) | 62 mm |