Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
BIOPHARMAGREENER_CHOICE
Product Code. p-4524822
Change view
Click to view available options
Volume (Metric):
150 mL
500 mL
1000 mL
Material (Membrane):
PES
SFCA
Pore Size:
0.2 μm
0.45 μm
Fits Bottle Neck Size (Metric):
33mm
45mm
Unit Size:
Pack of 12
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Material (Membrane) Pore Size Fits Bottle Neck Size (Metric) Unit Size
10441411 150 mL PES 0.2 μm 33mm Pack of 12
10598282 150 mL PES 0.2 μm 45mm Pack of 12
10743281 150 mL PES 0.45 μm 33mm Pack of 12
10319590 150 mL PES 0.45 μm 45mm Pack of 12
10526952 500 mL PES 0.2 μm 33mm Pack of 12
10300461 500 mL PES 0.2 μm 45mm Pack of 12
10706971 500 mL PES 0.45 μm 33mm Pack of 12
10646421 500 mL PES 0.45 μm 45mm Pack of 12
10491093 1000 mL PES 0.2 μm 33mm Pack of 12
10401103 1000 mL PES 0.2 μm 45mm Pack of 12
10352911 150 mL SFCA 0.2 μm 33mm Pack of 12
10139560 150 mL SFCA 0.2 μm 45mm Pack of 12
10027780 150 mL SFCA 0.45 μm 33mm Pack of 12
10624281 150 mL SFCA 0.45 μm 45mm Pack of 12
10037790 500 mL SFCA 0.2 μm 33mm Pack of 12
10201371 500 mL SFCA 0.2 μm 45mm Pack of 12
10529051 500 mL SFCA 0.45 μm 33mm Pack of 12
10549241 500 mL SFCA 0.45 μm 45mm Pack of 12
10139800 1000 mL SFCA 0.2 μm 33mm Pack of 12
10790592 1000 mL SFCA 0.2 μm 45mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Product Code. 10441411 Supplier Thermo Scientific™ Supplier No. 5963320
Add to basket
Add to basket
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Filter Type Membrane Filter
Material Polystyrene
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
Fits Bottle Neck Size (Metric) 33mm
Type Bottle Top Filter
Volume (Metric) 150 mL
Diameter (Metric) 50 mm
Diameter (Metric) Membrane 50 mm
Fit for Purpose Sterile
Show More Show Less
Can I remove the filter membranes from the Nalgene Rapid-Flow Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes?
No, the filter membranes for the bottle top filters are molded in and cannot be removed.