missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Heptyl)-n-octylamine, 98+%, Thermo Scientific™

Catalog Number p-7058495
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
26627-77-0
C15H33N
227.436
MFCD03093076
HAEBNPLWJOGFDQ-UHFFFAOYSA-N
n-n-heptyl-n-octylamine, heptyloctylamine, n-heptyloctylamine, heptyl-octyl-amine, n-heptyl-n-octylamine, heptyl octyl amine, 1-octanamine, n-heptyl
12905714
N-heptyloctan-1-amine
CCCCCCCCNCCCCCCC
This item is not returnable. View return policy
26627-77-0
C15H33N
227.436
MFCD03093076
HAEBNPLWJOGFDQ-UHFFFAOYSA-N
n-n-heptyl-n-octylamine, heptyloctylamine, n-heptyloctylamine, heptyl-octyl-amine, n-heptyl-n-octylamine, heptyl octyl amine, 1-octanamine, n-heptyl
12905714
N-heptyloctan-1-amine
CCCCCCCCNCCCCCCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.442
Quantity 250 mg
UN Number UN2735
Beilstein 2346655
Sensitivity Air Sensitive
Formula Weight 227.44
Percent Purity ≥98%
Chemical Name or Material N-(n-Heptyl)-n-octylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'