missing translation for 'onlineSavingsMsg'
Learn More

n-Butanol (Clear, Colorless Liq.), Fisher BioReagents

Catalog Number p-24917
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
71-36-3
C4H10O
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
This item is not returnable. View return policy
71-36-3
C4H10O
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
n-Butanol is used as an analytical reagent and solvent.
TRUSTED_SUSTAINABILITY
Melting Point -89°C
Color Colorless
Acidity 0.005% max.
Density 0.810g/cm³
Boiling Point 117.6°C
Quantity 25 mL
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥99.8 %
Linear Formula CH3CH2CH2CH2OH
Vapor Pressure 6.7mbar at 20°C
UN Number 1120
Viscosity 2.95 mPa/s at 20°C
Water 0.1% max.
Merck Index 15, 1542
Formula Weight 74.12
Percent Purity ≥99.8%
Chemical Name or Material n-Butanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'