missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Pyrrole, 98%, Thermo Scientific Chemicals

Catalog Number p-4323692
Click to view available options
Quantity:
25 g
100 g
5176-27-2
C9H13NO2
167.21
IZPYBIJFRFWRPR-UHFFFAOYSA-N
tert-butyl 1h-pyrrole-1-carboxylate, n-boc-pyrrole, n-t-boc-pyrrole, tert-butyl 1-pyrrolecarboxylate, t-butyl 1h-pyrrole-1-carboxylate, 1h-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl-1h-pyrrole, 1-boc pyrrole, 1-boc-pyrrole, acmc-209kvp
643494
tert-butyl pyrrole-1-carboxylate
CC(C)(C)OC(=O)N1C=CC=C1
5176-27-2
C9H13NO2
167.21
IZPYBIJFRFWRPR-UHFFFAOYSA-N
tert-butyl 1h-pyrrole-1-carboxylate, n-boc-pyrrole, n-t-boc-pyrrole, tert-butyl 1-pyrrolecarboxylate, t-butyl 1h-pyrrole-1-carboxylate, 1h-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl-1h-pyrrole, 1-boc pyrrole, 1-boc-pyrrole, acmc-209kvp
643494
tert-butyl pyrrole-1-carboxylate
CC(C)(C)OC(=O)N1C=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 97.0°C (2.3 Torr)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1
Formula Weight 167.21
Percent Purity 98%
Physical Form Solution
Chemical Name or Material N-BOC-Pyrrole
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-Pyrrole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'