Learn More
Macherey Nagel N 9 PP screw cap, red,center hole PTFE red/Silicone white/PTFE red
Brand:  Macherey Nagel 702084
N 9 PP screw cap, red,center hole PTFE red/Silicone white/PTFE red
Product Code. 12745232
  • 31.70€ / Each