Learn More
N-(4-Ethylbenzyl)-N-methylamine, Thermo Scientific™
Catalog Number p-4510121
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
568577-84-4
C10H15N
149.237
JCSDSVXBTRWEJS-UHFFFAOYSA-N
n-methyl-4-ethylbenzylamine, 1-4-ethylphenyl-n-methylmethanamine, n-4-ethylbenzyl-n-methylamine, 4-ethylphenyl methyl methyl amine, benzenemethanamine, 4-ethyl-n-methyl, 1-4-ethylphenyl-n-methyl-methanamine, 4-ethylbenzyl methylamine, 4-ethylphenyl methyl methylamine
2060575
1-(4-ethylphenyl)-N-methylmethanamine
CCC1=CC=C(C=C1)CNC
568577-84-4
C10H15N
149.237
JCSDSVXBTRWEJS-UHFFFAOYSA-N
n-methyl-4-ethylbenzylamine, 1-4-ethylphenyl-n-methylmethanamine, n-4-ethylbenzyl-n-methylamine, 4-ethylphenyl methyl methyl amine, benzenemethanamine, 4-ethyl-n-methyl, 1-4-ethylphenyl-n-methyl-methanamine, 4-ethylbenzyl methylamine, 4-ethylphenyl methyl methylamine
2060575
1-(4-ethylphenyl)-N-methylmethanamine
CCC1=CC=C(C=C1)CNC
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 149.24
Percent Purity 97%
Chemical Name or Material N-(4-Ethylbenzyl)-N-methylamine