Learn More
MP Biomedicals™ b-Lactamase [E.C. 3.5.2.6]
From Bacillus cereus
Brand: MP Biomedicals™ 0219003401
Additional Details : CAS : 9073-60-3 Weight : 0.00100kg
Description
Lyophilized solid. Activity: Each vial contains 500U β-Lactamase I and 50U β-Lactamase II. Unit Definition: One unit is defined as the amount of enzyme which will hydrolyze 1.0μm of benzyl penicillin and 1.0μm of cephalosporin C per minute at 25°C in the presence of EDTA pH 7.0.
Specifications
7 | |
4°C | |
1 Vial | |
32000 | |
Lyophilized Solid |
Destroy beta-Lactam antibiotics in body fluids or culture media and It is used in the production of Penicillin | |
Bacillus cereus | |
b-Lactamase | |
Clavulanic Acid, Sulbactam, Tazobactam |