Learn More
ACROBiosystems Mouse IL-10 Protein, His Tag
209.03€ - 457.25€
Specifications
Accession Number | NP_034678 |
---|---|
Molecular Weight (g/mol) | 19.6 kDa |
Source | Mouse IL-10, His Tag (IL0-M4248) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Ser 178 (Accession # NP_034678). |
Immunogen | Ser 19 - Ser 178 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15827017
|
ACROBiosystems
IL0-M4248-20UG |
20 μg |
209.03€
20µg |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
15837017
|
ACROBiosystems
IL0-M4248-50UG |
50 μg |
457.25€
50µg |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
Interleukin-10 (IL-10) is also known as human cytokine synthesis inhibitory factor (CSIF), is an anti-inflammatory cytokine. IL-10 is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes. Mature human IL-10 shares 72% - 86% amino acid sequence identity with bovine, canine, equine, feline, mouse, ovine, porcine, and rat IL-10. Whereas human IL-10 is active on mouse cells, mouse IL-10 does not act on human cells. IL-10 is capable of inhibiting synthesis of pro-inflammatory cytokines such as IFN-γ, IL-2, IL-3, TNFα and GM-CSF made by cells such as macrophages and regulatory T-cells. It also displays a potent ability to suppress the antigen-presentation capacity of antigen presenting cells. However, it is also stimulatory towards certain T cells and mast cells and stimulates B cell maturation and antibody production. Knockout studies suggested the function of Interleukin-10 / IL-10 as an essential immunoregulator in the intestinal tract. Patients with Crohn's disease react favorably towards treatment with bacteria producing recombinant interleukin-10, showing the importance of interleukin-10 for counteracting excessive immunity in the human body.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.6 kDa. The protein migrates as 21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_034678 | |
Mouse IL-10, His Tag (IL0-M4248) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Ser 178 (Accession # NP_034678). | |
-20°C | |
IL-10 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
19.6 kDa | |
Ser 19 - Ser 178 | |
CSIF,IL-10,IL10A,TGIF,B-TCGF,GVHDS,MGC126450,MGC126451,RP11-262N9.1,Interleukin-10 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |