Learn More
ACROBiosystems Mouse FOLR1 Protein, His Tag
Brand: ACROBiosystems FO1-M5225-100UG
Additional Details : Weight : 0.00001kg
Description
Folate Receptor 1 (FOLR1) is also known as Folate receptor alpha, Folate Binding Protein (FBP), FOLR, and is a member of the folate receptor (FOLR) family. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and mediate delivery of 5-methyltetrahydrofolate to the interior of cells. Mature FOLR1 is an N-glycosylated protein that is anchored to the cell surface by a GPI linkage. FOLR1 is predominantly expressed on epithelial cells and is dramatically upregulated on many carcinomas. FOLR1 is internalized to the endosomal system where it dissociates from its ligand before recycling to the cell surface. A soluble form of FOLR1 can be proteolytically shed from the cell surface into the serum and breast milk. Defects in FOLR1 are the cause of neurodegeneration due to cerebral folate transport deficiency (NCFTD). NCFTD is an autosomal recessive disorder resulting from brain-specific folate deficiency early in life.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 32-34 kDa and 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_032060.2 | |
100 μg | |
Thr 25 - Ser 232 | |
FOLR-1,FBP,FOLR | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
26.2 kDa | |
Mouse FOLR1, His Tag (FO1-M5225) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession # P35846-1). | |
-20°C | |
FOLR1 | |
Mouse | |
Tris and Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |