missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse BTLA Protein, Fc Tag

Brand:  ACROBiosystems BTA-M5253-1MG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15816077

  • 1815.83€ / 1mg


Estimated Shipment: 25-10-2021
Add to basket

Description

Description

B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.8 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by SDS-PAGE
Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession # Q32MV9).
-20°C
BTLA
Fc Tag
HEK293
Mouse
42.8 kDa
1 mg
Glu 30 - Gly 176
BTLA,CD272
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Videos
SDS
Documents
Certificates