missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF92, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ZNF92.
Brand: Abnova H00168374-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: FNQSSIFTKHKIIHTEGKSYKCEKCGNAFNQSSNLTARKIIYTGEKPYKYEECDKAFNKFSTLITHQIIYTGEKPCKHECGRAFNKSSNYTKEKLQTSpecifications
ZNF92 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ZNF92. | |
ZNF92 | |
HPF12/HTF12/TF12 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_152626 | |
ZNF92 | |
ZNF92 (NP_689839, 490 a.a. ∽ 586 a.a) partial recombinant protein with GST tag. | |
RUO | |
168374 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only