missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA135468
Additional Details : Weight : 0.01000kg
Description
This antibody can be used as a coating antibody in a Sandwich ELISA with Product # SA1-36099 (FITC). Reconstitute with 1 mL distilled water.
Human IgG (H+L) Monoclonal antibody specifically detects Human IgG (H+L) in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen).Specifications
Human IgG (H+L) | |
Monoclonal | |
0.5 mg/mL | |
0.01M sodium borate, PBS with 1% mannitol, 1.0% dextran and no preservative; pH 8 | |
Purified human IgG. | |
1 mL | |
Secondary | |
-20°C | |
IgG1 |
ELISA, Immunohistochemistry (Frozen) | |
EFE-565 | |
Unconjugated | |
Mouse | |
purified | |
RUO | |
Human | |
Lyophilized |