missing translation for 'onlineSavingsMsg'
Learn More

CD171 Mouse anti-Human, BUV395, Clone: 5G3, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 565736

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15824499

  • 565.00€ / Each


Estimated Shipment: 08-10-2021
Add to basket

Description

Description

The antibody was conjugated to BD Horizon BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

Specifications

Specifications

CD171
5G3
L1 Neurite Cell Adhesion Molecule; N-CAM-L1; L1CAM ; CAML1; MIC5
Affinity Purified
Human SK-N-AS Neuroblastoma Cell Line
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
Brilliant Ultraviolet 395
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG2a
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
SDS
Documents
Certificates

For Research Use Only.