Learn More
CD14 Mouse anti-Human, BUV805, Clone: M5E2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565779
Additional Details : Weight : 0.01000kg
Description
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
The antibody was conjugated to BD Horizon BUV805 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348nm and an acceptor dye with an Em Max at 805 nm. BD Horizon Brilliant BUV805 can be excited by the ultraviolet laser (355 nm) and detected with a 820/60 filter and a 770LP.
Specifications
CD14 | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CD14 Protein | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M5E2 | |
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
IgG2a κ |
For Research Use Only.