Learn More
Molecular Sieves, Grade 514 (Type 4A; 8-12 mesh), Fisher BioReagents
Brand: Fisher Bioreagents BP2631-500
Additional Details : CAS : 1344-00-9 Weight : 0.55000kg
Description
Used for dehydration of liquids.Specifications
8 to 12 Mesh | |
500 g | |
2.1g/cm³ | |
Molecular Sieves, Grade 514 | |
Solid | |
AlNaO6Si2 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N |
Beige | |
Pass Test | |
Glass Bottle | |
Type 4A; 8-12 Mesh | |
1318-02-1,14808-60-7 | |
Aluminum Sodium Silicate | |
202.14 |