missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MM 11253

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective RARγ antagonist. Blocks the growth inhibitory ability of RARγ-selective agonists in squamous cell carcinoma (SCC)-25 cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 462.67 |
| Purity | >98% |
| Molecular Weight (g/mol) | 462.666 |
| Chemical Name or Material | MM 11253 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction