Learn More
VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters
Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.
Brand: VistaLab™ Technologies 1108
Additional Details : Weight : 0.50000kg
Description
- “To deliver” capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Capacity engraved on bonnet
Specifications
±2% | |
Nozzle Insert, Calibration Key | |
1 Ea. | |
Limited Lifetime | |
Aluminum alloy and Stainless steel | |
1 |
Not Autoclavable | |
±0.53% | |
Non-sterile | |
Silver | |
MLA Precision | |
75 µL |