missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Minisart™ PES Venting Filter

Sartorius Stedim Biotech™ Minisart™ PES Venting Filter is a premium quality, hydrophobic PES filter designed for venting and gas filtration.
Brand: Sartorius Stedim Biotech™ 1757G----------Q
This item is not returnable.
View return policy
Description
- Premium filters for fail-safe filtration
- Can be used for sterile filtration
- Suitable for venting or filtration of gases and air
- Can be sterilized by gamma irradiation, autoclaving or EO

Specifications
White | |
PES | |
Non-sterile | |
Polypropylene | |
26 mm |
Hose Barb | |
0.2 μm | |
Venting and gas filtration | |
500/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction