Learn More
Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates
For use in flexibility in diagnostics. Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates are suitable for transmission measurements.
1100.00€ - 1125.00€
Specifications
Format | Strip Plate |
---|---|
Array | 8 x 12 |
Color | Clear |
Material | Polystyrene |
No. of Wells | 96 |
Product Code | Brand | Binding Property | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Binding Property | Price | Quantity & Availability | |||||
10033281
|
Greiner Bio-One
701070 |
Medium Binding |
1100.00€
Pack of 100 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10063291
|
Greiner Bio-One
701071 |
High Binding |
1125.00€
Pack of 100 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- For optical measurements
- Measuring light beam is not deflected by well profile
- Specifically treated to provide incresed protein binding
Specifications
Strip Plate | |
Clear | |
96 | |
Flat Bottom (F-well) |
8 x 12 | |
Polystyrene | |
Non-sterile | |
100/Cs. |