Learn More
Brand™ Micro Haematocrit Capillaries, Non-Heparinized
For use in haematocrit centrifuges.
Brand:  Brand™ 749321
Additional Details : Weight : 0.80000kg
Product Code. 11884040
  • 28.35€ / Pack of 1000
Estimated Shipment date
from Supplier 29-03-2019
Add to basket
Description
Description
  • Without calibration mark
  • Blue color code, for use with heparinized venous blood
  • CE-marked acc. to IVD-Directive 98/79 EC
  • Pack quantity of 1000; 10 glass boxes of 100
Specifications
Specifications
Blue
1.1 - 1.2mm (Outer: 1.5 - 1.6mm)
75 ± 0.5mm
With approx. 0.2mm wall thickness
Videos
Documentation