Learn More
Chemetrics™ Methylene Blue Method Sulfide VACUettes Refill
Brand: Chemetrics™ R9510D
Additional Details : Weight : 0.03000kg Transport : UN number : 1760 Chem class : 8 Pack group : II
Specifications
Sulfides are naturally present in ground waters as a result of leaching from sulfur-containing mineral deposits | |
30 Pack | |
Methylene Blue |
CHEMets Kit | |
0 to 30 & 30 to 300ppm | |
Sulfide VACUettes Refill |
Safety and Handling
- Sulfide Vacu-vials Ampoules and CHEMets & VACUettes Refills
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P234-Keep only in original container.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to authorised chemicallandfill or if organic to high temperature incineration.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid, isopropanol, N,N-dimethyl p-phenylenediamine oxalate