missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl benzenesulfinate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 1.1940g/mL |
Boiling Point | 110.0°C to 112.0°C (16.0 Torr) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Quantity | 5 mL |
Specific Gravity | 1.194 |
Formula Weight | 156.2 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Methyl benzenesulfinate, 98%
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'