Learn More
Methyle4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 181.19 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 4-formyl-2, 5-dimethyl-1H-pyrrole-3-carboxylate |