Learn More
Methyl 2,4-dichlorobenzoate, 97%, Thermo Scientific™
Catalog Number p-4228800
Click to view available options
Quantity:
5 g
25 g
35112-28-8
C8H6Cl2O2
205.04
MFCD00085117
VCRWILYAWSRHBN-UHFFFAOYSA-N
methyl2,4-dichlorobenzoate, 2,4-dichlorobenzoic acid methyl ester, benzoic acid, 2,4-dichloro-, methyl ester, methyl-2,4-dichlorobenzoate, pubchem3713, methyl ester of 2,4-dichlorobenzoic acid, acmc-209id8, ksc497m5b, methyl 2,4-dichloro benzoate
37055
methyl 2,4-dichlorobenzoate
COC(=O)C1=C(C=C(C=C1)Cl)Cl
35112-28-8
C8H6Cl2O2
205.04
MFCD00085117
VCRWILYAWSRHBN-UHFFFAOYSA-N
methyl2,4-dichlorobenzoate, 2,4-dichlorobenzoic acid methyl ester, benzoic acid, 2,4-dichloro-, methyl ester, methyl-2,4-dichlorobenzoate, pubchem3713, methyl ester of 2,4-dichlorobenzoic acid, acmc-209id8, ksc497m5b, methyl 2,4-dichloro benzoate
37055
methyl 2,4-dichlorobenzoate
COC(=O)C1=C(C=C(C=C1)Cl)Cl
Color Colorless to Yellow
Boiling Point 132.0°C (15.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 205.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 2,4-dichlorobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2,4-dichlorobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.