Learn More
Methylcyclopentadiene dimer, 90%, stabilized, Thermo Scientific Chemicals
Catalog Number p-3758372
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
26472-00-4
C12H16
160.26
MFCD00082297
26472-00-4
C12H16
160.26
MFCD00082297
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -51.0°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 200.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Refractive Index 1.4975 to 1.4995
Quantity 250 g
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene and ether
Formula Weight 160.26
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methylcyclopentadiene dimer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methylcyclopentadiene dimer
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.