missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (S)-(+)-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Methyl (S)-(+)-3-hydroxybutyrate serves as an optically active starting material used in organic synthesis. It is also used in the preparation of (-)-methyl elenolate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl (S)-(+)-3-hydroxybutyrate serves as an optically active starting material used in organic synthesis. It is also used in the preparation of (-)-methyl elenolate.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with oxidizing agents, bases and acids.
Methyl (S)-(+)-3-hydroxybutyrate serves as an optically active starting material used in organic synthesis. It is also used in the preparation of (-)-methyl elenolate.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with oxidizing agents, bases and acids.
Specifications
Density | 1.055 |
Boiling Point | 56°C to 58°C (11 mmHg) |
Flash Point | 73°C (163°F) |
Refractive Index | 1.422 |
Quantity | 1 g |
Beilstein | 1720583 |
Solubility Information | Miscible with alcohol. Immiscible with water. |
Optical Rotation | +48.5° (c=1.3 in Chloroform) |
Formula Weight | 118.13 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'