missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-L-arabinopyranoside, 98+%, Thermo Scientific Chemicals

Catalog Number p-7047782
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1825-00-9
C6H12O5
164.157
MFCD00047547
ZBDGHWFPLXXWRD-AZGQCCRYSA-N
methyl beta-l-arabinopyranoside, 2s,3r,4s,5s-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s-2-methoxyoxane-3,4,5-triol, methyl-b-l-arabinopyranoside, methyl, a-l-arabinopyranoside, methyl-beta-l-arabinopyranoside, .beta.-l-arabinopyranoside, methyl, methyl .beta.-l-arabinopyranoside, .beta.-l-arabinopyranose methyl glycoside, arabinopyranoside, methyl, .beta.-l
102169
(2S,3R,4S,5S)-2-methoxyoxane-3,4,5-triol
COC1C(C(C(CO1)O)O)O
This item is not returnable. View return policy
1825-00-9
C6H12O5
164.157
MFCD00047547
ZBDGHWFPLXXWRD-AZGQCCRYSA-N
methyl beta-l-arabinopyranoside, 2s,3r,4s,5s-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s-2-methoxyoxane-3,4,5-triol, methyl-b-l-arabinopyranoside, methyl, a-l-arabinopyranoside, methyl-beta-l-arabinopyranoside, .beta.-l-arabinopyranoside, methyl, methyl .beta.-l-arabinopyranoside, .beta.-l-arabinopyranose methyl glycoside, arabinopyranoside, methyl, .beta.-l
102169
(2S,3R,4S,5S)-2-methoxyoxane-3,4,5-triol
COC1C(C(C(CO1)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 167°C to 172°C
Quantity 1 g
Beilstein 4658388
Optical Rotation +243° (c=2 in Water)
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material Methyl beta-L-arabinopyranoside

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'