missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl beta-D-ribofuranoside, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography.
Solubility
Soluble in water
Notes
Store at 2-8°C. Incompatible with heat, oxidizing agents.
D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography.
Solubility
Soluble in water
Notes
Store at 2-8°C. Incompatible with heat, oxidizing agents.
Specifications
Melting Point | 78°C |
Color | White |
Quantity | 5 g |
Solubility Information | Soluble in water |
Optical Rotation | −50° (c=2 in Water) |
Formula Weight | 164.16 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Methyl beta-D-ribofuranoside |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'