missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-galactopyranoside monohydrate, Thermo Scientific™

Catalog Number p-7044748
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
34004-14-3
C7H14O6
194.18
MFCD00150748
HOVAGTYPODGVJG-PZRMXXKTSA-N
methyl alpha-d-galactopyranoside monohydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol hydrate, methyl-alpha-d-galactopyranoside, methyl ?-galactoside hydrate, methyl a-d-galactopyranoside,monohydrate, methyl alpha-d-galactopyranoside hydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxyoxane-3,4,5-triol hydrate
2724703
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol;hydrate
CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
This item is not returnable. View return policy
34004-14-3
C7H14O6
194.18
MFCD00150748
HOVAGTYPODGVJG-PZRMXXKTSA-N
methyl alpha-d-galactopyranoside monohydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol hydrate, methyl-alpha-d-galactopyranoside, methyl ?-galactoside hydrate, methyl a-d-galactopyranoside,monohydrate, methyl alpha-d-galactopyranoside hydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxyoxane-3,4,5-triol hydrate
2724703
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol;hydrate
CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl alpha-D-galactopyranoside monohydrate is used in medicine, industry.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 105°C to 108°C
Quantity 1 g
Beilstein 81570
Solubility Information Insoluble in water.
Optical Rotation +169° (c=1.5 in Methanol)
Formula Weight 212.20 (194.18 Anhydrous)
Chemical Name or Material Methyl alpha-D-galactopyranoside monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'