missing translation for 'onlineSavingsMsg'
Learn More

Valeraldehyde-DNPH in Acetonitrile 100μg/mL, Fisher Chemical™

Catalog Number 12952964
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 100μg/mL
CAS 2057-84-3
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C11H14N4O4
UN Number 1648
InChI Key XGVOZGLOQUHZBZ-KPKJPENVSA-N
SMILES CCCCC=NNC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2,4-dinitro-N-[(E)-pentylideneamino]aniline
Molecular Weight (g/mol) 266.257
PubChem CID 9601271
Solvent or Matrix Acetonitrile
Physical Form Liquid
Chemical Name or Material Valeraldehyde-DNPH
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Valeraldehyde-DNPH 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'