missing translation for 'onlineSavingsMsg'
Learn More

Mercury Standard Solution (Plasma HIQU), Chem Lab™

Catalog Number 11969082
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Concentration 1000μg/mL
Quantity 100 mL
compliance-icons
Product Identifier
  • Mercury standard solution 1000
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid 69%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'