Learn More
Masterflex™ Pump Head Adapter
Connect any I/P™ pump head to a 56C frame motor
1195.00€ - 1450.00€
Product Code | Brand | Flow Rate | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Price | Quantity & Availability | |||||
11855191
|
Masterflex™
77495-00 |
0.06 to 3400 mL/min. |
1195.00€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11856733
|
Masterflex™
77490-00 |
0.12 to 13 L/min. |
1450.00€
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Description
- Cast aluminum adapters feature built-in gear box for easy setup
- Compatible with all I/P pump heads
- Suitable for use with 1⁄10 hp to 1⁄2 hp 56C frame motor and 650 rpm max pump head speed
- Flow rate 0.12 to 19 LPM with I/P tubing and 3.1:1 gear ratio—flow rate depends on drive rpm, pump head, and tubing size
Specifications
1 Each | |
Pump Head Adapter |