Learn More
Masterflex™ L/S™ Peroxide-Cured Silicone Spooled Pump Tubing
Excellent biocompatibility for a wide range of applications
725.00€ - 1325.00€
Specifications
Product Type | Pump Tubing |
---|---|
Autoclavable | Yes |
Overall Length (English) | 250 ft. |
Overall Length (Metric) | 76.2 m |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11880130
|
Masterflex™
96407-15 |
15 |
1200.00€
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
13296980
|
Masterflex™
96407-14 |
14 |
725.00€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11830110
|
Masterflex™
MFLX96407-24 |
24 |
1260.00€
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
13133828
|
Masterflex™
96407-16 |
16 |
805.00€
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
13143828
|
Masterflex™
96407-17 |
17 |
1325.00€
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Greater physical compression capability
- Pharmaceutical, biotech, peristaltic pumping, and food applications
- Excellent heat resistance, low compression set, odorless, nontoxic, translucent
- Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Specifications
Pump Tubing | |
Yes | |
250 ft. | |
76.2 m |