Learn More
Tocris Bioscience™ Masitinib
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Masitinib is a potent inhibitor of the tyrosine kinase receptor colony-stimulating factor 1R (CSF-1R) (IC50 = 90 nM). It is also an NLRP3 inflammasome activator. Masitinib additionally inhibits the proteolytic activity of SARS-CoV-2 main protease (Mpro) and blocks viral replication, as well as acting as a PDGFRα/β inhibitor. In vivo, Masitinib ameliorates cognitive impairment in transgenic mice model of Alzheimer's disease, and slows disease progression in a rat model of amyotrophic lateral sclerosis (ALS). Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-thiazolyl]amino]phenyl]benzamide |
| CAS | 790299-79-5 |
| Quantity | 10 mg |
| Target | CD117/c-kit |
| Molecular Formula | C28H30N6OS |
| Purity | 0.98 |