missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAP3K3 (Human) Recombinant Protein
Human MAP3K3 full-length ORF ( AAH10464, 1 a.a. - 90 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH10464 |
---|---|
Gene ID (Entrez) | 4215 |
Name | mitogen-activated protein kinase kinase kinase 3 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16157741
|
Abnova™
H00004215-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16167741
|
Abnova™
H00004215-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Beschreibung
- Sequence: MNEANVMLPYSGKEEPVLPVAMTLPLPGRGPRCGTAATEGGSSFVNAVVSVLQVGVTLMLYPVSKLETVCALWALSTPALGLGLGCIEKS
Spezifikation
AAH10464 | |
mitogen-activated protein kinase kinase kinase 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
MAPKKK3, MEKK3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4215 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAP3K3 | |
GST |