missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Manganese(Iv) Oxide, activated, 85%, Honeywell™ Fluka™

Activated, ~85%, <10 μm

Brand:  Honeywell Fluka™ 217646-100G

Additional Details : CAS : 1313-13-9 Weight : 0.10000kg

Packaging Poly Bottle
Quantity 100g

 View more versions of this product

Product Code. 15622070

  • 29.77€ / 100g


EU Stock 5
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1313-13-9
86.936
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese

Specifications

Manganese(Iv) Oxide
MnO2
NONH for all modes of transport
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
85%
Poly Bottle
1313-13-9
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
O=[Mn]=O
86.936
86.94g/mol
Activated
100g
Videos
SDS Documents