missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Manganese(IV) Oxide, ≥90%, Reagent grade, Honeywell™

10 μm, Reagent Grade,≥90%

Brand:  Honeywell™ 310700-2.5KG

Additional Details : CAS : 1313-13-9 Weight : 2.50000kg

Packaging Poly Bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 15623210

  • 331.35€ / 2.50kg


Estimated Shipment date
from Supplier 21-06-2019
Add to basket

Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

1313-13-9
86.936
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese

Specifications

Manganese(Iv) Oxide
MnO2
NONH for all modes of transport
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
≥90%
5.21g/cm3
535°C
1313-13-9
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
O=[Mn]=O
86.936
86.94g/mol
Reagent
Poly Bottle
2.5kg
Videos
SDS