Learn More
Thermo Scientific™ Imject™ Maleimide-Activated Blue Carrier™ Protein
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
123.00€ - 1084.00€
Specifications
Content And Storage | Upon receipt store at -20°C. Product is shipped on ice. |
---|---|
Description | Imject Maleimide-Activated Blue Carrier Protein |
Product Line | Blue Carrier™, Imject™ |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11821184
|
Thermo Scientific™
77661 |
Imject Maleimide-Activated Blue Carrier Protein | 2 mg |
123.00€
2mg |
In Stock
Log in to see stock. |
||||
11899933
|
Thermo Scientific™
77662 |
Imject Maleimide-Activated Blue Carrier Protein | 10 x 10 mg |
1084.00€
Pack of 10 |
Estimated Shipment: 11-04-2024
Log in to see stock. |
||||
11810094
|
Thermo Scientific™
77664 |
Imject Maleimide-Activated Blue Carrier Protein | 10 mg |
372.00€
10mg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
||||
Description
Thermo Scientific™ Maleimide-Activated Blue Carrier Protein is sulfhydryl-reactive immunogenic hemocyanin (CCH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Blue Carrier Protein is a large homocyanin that is highly immunogenic like keyhole limpet hemocyanin (KLH) yet is more soluble. The carrier is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each Blue Carrier Protein molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest.
Highlights:
Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of carrier, ensuring consistently high levels of conjugatio
Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
High-quality carrier protein – Blue Carrier Protein is ultrapure Concholepas concholepas hemocyanin (CCH) prepared from molluscan hemolymph
Highly immunogenic and soluble – CCH has a high molecular mass that elicits a stronger immune response than BSA or ovalbumin
Specifications
Upon receipt store at -20°C. Product is shipped on ice. | |
Imject Maleimide-Activated Blue Carrier Protein | |
Blue Carrier™, Imject™ |
For Research Use Only. Not for use in diagnostic procedures.