Learn More
Gilson™ Maintenance Kit for Pipetman™
Manufactured by Gilson™. Gilson™ Maintenance Kit for Pipetman™ are guaranteed to deliver value and assure reliability and precision.
32.64€ - 53.65€
Specifications
Includes 1 Tip-holder, 3 piston seals and 3 O-rings
View More Specs
Products 6
Product Code Brand For Use With (Application) Price Quantity & Availability  
Product Code Brand For Use With (Application) Price Quantity & Availability  
10377132 View Documents Gilson™
F144502
For Pipetman: P10 53.65€
Each View alternative products
Save Up to 
»
In Stock 3
Add to basket
10737604 View Documents Gilson™
F144496
For Pipetman: P100/F25-F100 32.64€
Each View alternative products
Save Up to 
»
Estimated Shipment
20-01-2020
Add to basket
10757984 View Documents Gilson™
F144498
For Pipetman: P1000/F250-F1000 32.96€
Each View alternative products
Save Up to 
»
Estimated Shipment
20-01-2020
Add to basket
10133192 View Documents Gilson™
F144501
For Pipetman: P2/P2G 47.94€
Each View alternative products
Save Up to 
»
In Stock 1
Add to basket
10591273 View Documents Gilson™
F144497
For Pipetman: P200/F200 33.62€
Each View alternative products
Save Up to 
»
In Stock 4
Add to basket
10561103 View Documents Gilson™
F144499
For Pipetman™ P5000/F5000 43.26€
Each View alternative products
Save Up to 
»
Estimated Shipment
20-01-2020
Add to basket
Description
Description
  • Maintenance kit  
Specifications
Specifications
1 Tip-holder, 3 piston seals and 3 O-rings