missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, ≥99.5%, ReagentPlus™, Honeywell

Catalog Number p-7072299
Click to view available options
Quantity:
500 g
1 kg
2 kg
12 kg
7487-88-9
MgO4S
120.36
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
[Mg++].[O-]S([O-])(=O)=O
7487-88-9
MgO4S
120.36
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
[Mg++].[O-]S([O-])(=O)=O

Reagent Grade, anhydrous,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Quantity 500 g
UN Number NONH for all modes of transport
IUPAC Name magnesium;sulfate
Formula Weight 120.37g/mol
Percent Purity ≥99.5%
Vapor Pressure 0.1 mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'