missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide, puriss p.a, ≥97% (calcined substance, KT), Honeywell Fluka™

Catalog Number p-7120536
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]

Puriss. p.a., ACS Reagent,≥97% (calcined substance, KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 3600°C
Melting Point 2852°C
Color White
pH Alkaline
Physical Form Powder
Quantity 100 g
Linear Formula MgO
UN Number NONH for all modes of transport
Formula Weight 40.3g/mol
Percent Purity ≥97% (calcined substance, KT)
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Magnesium oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'